missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-bis-O-(1-Methylethylidene)-1-chlorosulfate beta-D-Fructopyranose (Technical Grade), TRC

Product Code. 30476338
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
Product Code. Quantity unitSize
30476338 500 mg 500mg
30466578 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 30476338

Brand: Toronto Research Chemicals TRCM304875500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150609-95-3
Formula Weight 358.0489
InChI Formula InChI=1S/C12H19ClO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3/t7-,8-,9+,12+/m1/s1
IUPAC Name (3aS,5aR,8aR,8bS)-3a-(chlorosulfonyloxymethyl)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a:3',5'-c]pyran
Molecular Formula C12 H19 Cl O8 S
Molecular Weight (g/mol) 358.79
SMILES CC1(C)O[C@@H]2CO[C@@]3(COS(=O)(=O)Cl)OC(C)(C)O[C@H]3[C@@H]2O1
Synonym 2,3:4,5-bis-O-(1-Methylethylidene)-β-D-fructopyranose 1-Chlorosulfate, β-D-Fructopyranose, 2,3:4,5-bis-O-(1-methylethylidene)-, chlorosulfate (9CI), 5H-Bis[1,3]dioxolo[4,5-b:4',5'-d]pyran, β-D-fructopyranose deriv., 2,3:4,5-Bis-O-(1-methylethylidene)-β-D-fructopyranose sulfonyl chloride
Chemical Name or Material 2,3:4,5-bis-O-(1-Methylethylidene)-1-chlorosulfate beta-D-Fructopyranose

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.