missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenylacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11465886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465886 5 g 5g
11455886 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465886 Supplier Thermo Scientific Alfa Aesar Supplier No. B22239.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 653-30-5 | Molecular Formula: C8H2F5N | Molecular Weight: 207.103 g/mol

2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-30-5
Molecular Formula C8H2F5N
Molecular Weight (g/mol) 207.103
MDL Number MFCD00013814
InChI Key YDNOJUAQBFXZCR-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile
PubChem CID 69554
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
SMILES C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Density 1.61
Boiling Point 110°C to 111°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.439
Quantity 5 g
UN Number UN3276
Beilstein 1882341
Solubility Information Insoluble in water.
Formula Weight 207.1
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.