missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenylacetonitrile, 96%, Thermo Scientific™

Product Code. 10145940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10145940 25 g Glass bottle 25g
10106973 1 g Glass bottle 1g
10531901 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10145940 Supplier Thermo Scientific Acros Supplier No. 296160250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-30-5
Molecular Formula C8H2F5N
Molecular Weight (g/mol) 207.103
MDL Number MFCD00013814
InChI Key YDNOJUAQBFXZCR-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile
PubChem CID 69554
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
SMILES C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6100g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula C6F5CH2CN
Refractive Index 1.4390 to 1.4410 (20°C, 589nm)
Quantity 25 g
Formula Weight 207.1
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.