missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzyl alcohol, 98%

Product Code. 11410063
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410063 100 g 100g
11490053 5 g 5g
11400063 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410063 Supplier Thermo Scientific Alfa Aesar Supplier No. A12119.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 440-60-8 | C7H3F5O | 198.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 440-60-8
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00004602
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
PubChem CID 9923
ChEBI CHEBI:44903
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O

Specifications

Melting Point 32°C to 36°C
Boiling Point 182°C
Flash Point 87°C (188°F)
Quantity 100 g
Beilstein 2052669
Formula Weight 198.09
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.