missing translation for 'onlineSavingsMsg'
Learn More

2',3',4',5',6'-Pentafluoroacetophenone, 97%

Product Code. 11381497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381497 1 g 1g
11391497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381497 Supplier Thermo Scientific Alfa Aesar Supplier No. L02063.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 652-29-9 | C8H3F5O | 210.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-29-9
Molecular Formula C8H3F5O
Molecular Weight (g/mol) 210.10
MDL Number MFCD00000296
InChI Key FBGHCYZBCMDEOX-UHFFFAOYSA-N
Synonym 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone
PubChem CID 69546
SMILES CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.476
Boiling Point 130°C to 131°C
Flash Point 65°C (149°F)
Refractive Index 1.4366
Quantity 1 g
Beilstein 2053225
IUPAC Name 1-(2,3,4,5,6-pentafluorophenyl)ethanone
Formula Weight 210.1
Percent Purity 97%
Chemical Name or Material 2',3',4',5',6'-Pentafluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.