missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethyl-1-butene, 99+%

Product Code. 10041620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041620 25 g Glass bottle 25g
10177403 1 g Glass bottle 1g
10726171 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10041620 Supplier Thermo Scientific Acros Supplier No. 140130250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-56-9 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-56-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00008850
InChI Key AUYRUAVCWOAHQN-UHFFFAOYSA-N
Synonym 2,3,3-trimethyl-1-butene, 1-butene, 2,3,3-trimethyl, triptene, acmc-209mdj, 2,3-trimethyl-1-butene, 1-butene,3,3-trimethyl, 1-butene,2,3,3-trimethyl, ch3 3cc ch3 =ch2
PubChem CID 11669
IUPAC Name 2,3,3-trimethylbut-1-ene
SMILES CC(=C)C(C)(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.7000g/mL
Boiling Point 78°C to 80°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3)3CC(CH3)=CH2
Refractive Index 1.4015 to 1.4035
Quantity 25 g
Beilstein 01, 221
Specific Gravity 0.7
Formula Weight 98.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 3, 3-Trimethyl-1-butene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,3-Trimethyl-1-butene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.