missing translation for 'onlineSavingsMsg'
Learn More

2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride, Thermo Scientific Chemicals

Product Code. 15400417
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400417 10 mg 10mg
15490407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400417 Supplier Thermo Scientific Alfa Aesar Supplier No. J64981.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 418788-90-6
Molecular Formula C11H16Cl2NO2
Molecular Weight (g/mol) 265.15
MDL Number MFCD06410993
InChI Key QVEIRCZEBQRCTR-UHFFFAOYSA-O
Synonym 2,3-dcpe hydrochloride, 2-3-2,3-dichlorophenoxy propyl amino ethanol hydrochloride, c11h15cl2no2.hcl, 2-3-2,3-dichlorophenoxy propylamino ethanol hydrochloride
PubChem CID 52913859
IUPAC Name 2-[3-(2,3-dichlorophenoxy)propylamino]ethanol;hydrochloride
SMILES OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl

Specifications

Color White
Quantity 10 mg
Solubility Information Soluble in water at ∽25mg/ml; Also soluble in ethanol,DMSO or DMF at ∽20mg/ml; Soluble in PBS,pH 7;2 at ∽10mg/ml.
Formula Weight 300.61
Physical Form Powder
Chemical Name or Material 2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.