missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Trityl-2H-tetrazol-5-yl)phenylboronic Acid, TRC

Product Code. 30450475
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450475 25 mg -
30452760 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30450475 Supplier Toronto Research Chemicals Supplier No. TRCT89000025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143722-25-2
Molecular Formula C26H21BN4O2
Molecular Weight (g/mol) 432.28
Synonym B-[2-[2-(Triphenylmethyl)-2H-tetrazol-5-yl]phenyl]boronic Acid, [2-[2-(Triphenylmethyl)-2H-tetrazol-5-yl]phenyl]boronic Acid
IUPAC Name [2-(2-trityltetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)c1ccccc1c2nnn(n2)C(c3ccccc3)(c4ccccc4)c5ccccc5

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H21BN4O2/c32-27(33)24-19-11-10-18-23(24)25-28-30-31(29-25)26(20-12-4-1-5-13-20,21-14-6-2-7-15-21)22-16-8-3-9-17-22/h1-19,32-33H
Formula Weight 432.1758
Chemical Name or Material 2-(2-Trityl-2H-tetrazol-5-yl)phenylboronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.