missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thiodiethanethiol, tech. 85%

Product Code. 11465986
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465986 250 g 250g
11445986 10 g 10g
11455986 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465986 Supplier Thermo Scientific Alfa Aesar Supplier No. B22304.30
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol

2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
SMILES SCCSCCS

Specifications

Density 1.183
Boiling Point 135°C to 136°C (10 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.5955
Quantity 250 g
Solubility Information Difficult to mix.
IUPAC Name 2-(2-sulfanylethylsulfanyl)ethanethiol
Formula Weight 154.32
Percent Purity ≥90%
Chemical Name or Material 2,2'-Thiodiethanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.