missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienylsulfonyl)ethanethioamide, 97%, Thermo Scientific™

Product Code. 10406662
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10406662 250 mg Amber Glass Bottle 250mg
10099151 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406662 Supplier Thermo Scientific Maybridge Supplier No. KM03009CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-34-3
Molecular Formula C6H7NO2S3
Molecular Weight (g/mol) 221.307
MDL Number MFCD00052277
InChI Key YNPIRUTXXLFXRU-UHFFFAOYSA-N
Synonym 2-2-thienylsulfonyl ethanethioamide, 2-thiophene-2-sulfonyl ethanethioamide, ethanethioamide,2-2-thienylsulfonyl, maybridge1_004617, 2-2-thienyl sulfonyl thioacetamide, 2-thiophen-2-ylsulfonyl ethanethioamide, 2-thien-2-yl sulphonyl ethanethioamide, 2-2-amino-2-thioxoethyl sulphonyl thiophene, 2-amino-1-2-thienylsulfonyl ethane-2-thione
PubChem CID 2778709
IUPAC Name 2-thiophen-2-ylsulfonylethanethioamide
SMILES C1=CSC(=C1)S(=O)(=O)CC(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 221.32
Percent Purity 97%
Chemical Name or Material 2-(2-Thienylsulfonyl)ethanethioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.