missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™

Product Code. 10713263
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10713263 250 mg 250mg
10580353 1 g 1g
10737305 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10713263 Supplier Thermo Scientific Maybridge Supplier No. RF05240CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58749-51-2
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.199
InChI Key KAJZMNLREUEZSH-UHFFFAOYSA-N
PubChem CID 2782179
IUPAC Name 2-(2-isocyanatoethyl)thiophene
SMILES C1=CSC(=C1)CCN=C=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym 2-2-isocyanatoethyl thiophene, 2-2-thienyl ethyl isocyanate, 2-thien-2-yl ethyl isocyanate, thiophene,2-2-isocyanatoethyl, 2-2-isocyanato-ethyl-thiophene, 2-2-thienyl ethanisocyanate, 2-2-isocyanatoethyl-thiophene, thiophene, 2-2-isocyanatoethyl, 2-thien-2-yl ethylisocyanate
Formula Weight 153.2
Percent Purity 95%
Chemical Name or Material 2-(2-Thienyl)ethyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.