missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)ethanol, 98%

Product Code. 10314552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314552 100 g Glass bottle 100g
10712263 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314552 Supplier Thermo Scientific Acros Supplier No. 366541000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5402-55-1 | C6H8OS | 128.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5402-55-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00005462
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
PubChem CID 79400
IUPAC Name 2-thiophen-2-ylethanol
SMILES OCCC1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown or Gray-Green to Yellow
Density 1.1530g/mL
Boiling Point 108°C (13.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5490 to 1.552
Quantity 100 g
Beilstein 17, V,3, 379
Specific Gravity 1.153
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in mek,alcohol,organic halogen solvents
Formula Weight 128.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Thienyl)ethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Thienyl)ethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.