missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)benzimidazole, 97%, Thermo Scientific™

Product Code. 15414648
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3878-18-0
Molecular Formula C11H8N2S
Molecular Weight (g/mol) 200.259
MDL Number MFCD00179920
InChI Key XXCGVPNWMZKOER-UHFFFAOYSA-N
Synonym 2-2-thienyl-1h-benzimidazole, 2-thiophen-2-yl-1h-benzoimidazole, 1h-benzimidazole, 2-2-thienyl, 2-thiophen-2-yl-1h-benzo d imidazole, 2-thiophen-2-yl-1h-benzimidazole, 2-thiophen-2-yl-1h-1,3-benzodiazole, 2-benzimidazol-2-ylthiophene, cambridge id 5793899, 2-2-thienyl benzimidazole, 2-2'-thienyl benzimidazole
PubChem CID 506600
IUPAC Name 2-thiophen-2-yl-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC=CS3

Specifications

Melting Point 230°C (decomposition)
Color Yellow to Brown
Quantity 1 g
Formula Weight 200.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(2-Thienyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'