missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Thenil, 98%

Product Code. 11476456
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476456 5 g 5g
11466456 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476456 Supplier Thermo Scientific Alfa Aesar Supplier No. B24753.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7333-07-5 | C10H6O2S2 | 222.28 g/mol

Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7333-07-5
Molecular Formula C10H6O2S2
Molecular Weight (g/mol) 222.28
MDL Number MFCD00173678
InChI Key UNWKVSDABPCZMK-UHFFFAOYSA-N
Synonym 2,2'-thenil, 1,2-di thiophen-2-yl ethane-1,2-dione, bis thiophen-2-yl ethane-1,2-dione, ethanedione, di-2-thienyl, 1,2-di 2-thienyl-1,2-ethanedione, 1,2-di 2-thienyl ethane-1,2-dione, 1,2-di-thiophen-2-yl-ethane-1,2-dione, 5-alpha-thenil, dithienylethanedione, 2,2'-bithenoyl
PubChem CID 275005
IUPAC Name 1,2-dithiophen-2-ylethane-1,2-dione
SMILES C1=CSC(=C1)C(=O)C(=O)C2=CC=CS2

Specifications

Melting Point 81°C to 84°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 222.29
Percent Purity 98%
Chemical Name or Material 2,2'-Thenil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.