Learn More
2,2'-Thenil, 98%
Description
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 81°C to 84°C |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 222.29 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,2'-Thenil |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.