missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzimidazole, 99%

Product Code. 10032090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032090 5 g Glass bottle 5g
10295880 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032090 Supplier Thermo Scientific Acros Supplier No. 167950050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1137-68-4 | C12H9N3 | 195.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-68-4
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00005586
InChI Key YNFBMDWHEHETJW-UHFFFAOYSA-N
Synonym 2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
PubChem CID 70821
IUPAC Name 2-pyridin-2-yl-1H-benzimidazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 220.0°C to 225.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 195.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-(2-Pyridyl)benzimidazole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.