missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Pyridyl)benzimidazole, 98+%

Product Code. 11314574
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314574 5 g 5g
11304574 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314574 Supplier Thermo Scientific Alfa Aesar Supplier No. L01339.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1137-68-4 | C12H9N3 | 195.23 g/mol

2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Pyridyl)benzimidazole is used as a chelating fluorescent ligand in the synthesis of cobalt(II) complex. It was also used in the preparation of (H(2)L(1))(2)[Ru(III)Cl(4)(CH(3)CN)(2)](2)[Ru(IV)Cl(4)(CH(3)CN)(2)]·2Cl·6H(2)O complex.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-68-4
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.23
MDL Number MFCD00005586
InChI Key YNFBMDWHEHETJW-UHFFFAOYSA-N
Synonym 2-2-pyridyl benzimidazole, 2-pyridin-2-yl-1h-benzo d imidazole, 2-pyridin-2-yl-1h-benzoimidazole, 2-pyridin-2-yl-1h-1,3-benzodiazole, 2-pyridin-2-yl-1h-benzimidazole, chembl72683, 1h-benzimidazole, 2-pyridinyl, 1h-benzimidazole, 2-2-pyridinyl, acmc-20aime, 2 2-pyridyl benzimidazole
PubChem CID 70821
IUPAC Name 2-pyridin-2-yl-1H-benzimidazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=N1

Specifications

Melting Point 221°C to 225°C
Quantity 5 g
Beilstein 151411
Solubility Information Insoluble in water.
Formula Weight 195.22
Percent Purity ≥98%
Chemical Name or Material 2-(2-Pyridyl)benzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.