missing translation for 'onlineSavingsMsg'
Learn More

[2.2]Paracyclophane, 99%

Product Code. 11400671
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400671 1 g 1g
11410671 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400671 Supplier Thermo Scientific Alfa Aesar Supplier No. B20136.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1633-22-3 | C16H16 | 208.304 g/mol

2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-22-3
Molecular Formula C16H16
Molecular Weight (g/mol) 208.304
MDL Number MFCD00003707
InChI Key OOLUVSIJOMLOCB-UHFFFAOYSA-N
Synonym 2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
PubChem CID 74210
SMILES C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2

Specifications

Melting Point 283°C to 288°C
Odor Odorless
Quantity 1 g
Beilstein 1910888
Solubility Information Insoluble in water.
Formula Weight 208.3
Percent Purity 99%
Chemical Name or Material [2.2]Paracyclophane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.