missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Oxydiethanol, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15598814 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15598814 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598814 Supplier Avantor J.T.Baker Supplier No. S85607

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 111-46-6
Flash Point 124.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C4H10O3
MDL Number MFCD00002882
Quantity 500 mL
Specific Gravity 1L = 1.12kg
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
SMILES OCCOCCO
IUPAC Name 2-(2-hydroxyethoxy)ethanol
Molecular Weight (g/mol) 106.12
PubChem CID 8117
ChEBI CHEBI:46807
Formula Weight 106.12
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material 2,2'-Oxydiethanol
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.