missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Oxybis(ethylamine), 98%

Product Code. 10235671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235671 1 g Glass bottle 1g
11925281 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235671 Supplier Thermo Scientific Acros Supplier No. 275840010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2752-17-2 | C4H12N2O | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2752-17-2
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00466591
InChI Key GXVUZYLYWKWJIM-UHFFFAOYSA-N
Synonym 2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine
PubChem CID 75982
IUPAC Name 2-(2-aminoethoxy)ethanamine
SMILES NCCOCCN

Specifications

Color Colorless
Density 0.9610g/mL
Boiling Point 69.0°C to 71.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2CH2OCH2CH2NH2
Refractive Index 1.4550 to 1.4580
Quantity 1 g
Beilstein 04,IV,1414
Specific Gravity 0.961
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 104.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 2'-Oxybis(ethylamine)
Show More Show Less
compliance-icons
Product Identifier
  • 2,2``-Oxybis(ethylamine)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.