missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar H66633.03

Quantity 1g

 View more versions of this product

Product Code. 15430107

  • £168.00 / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

104206-65-7
329.231
OUBCNLGXQFSTLU-UHFFFAOYSA-N
115355
2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
C14H10F3NO5
MFCD01752192
nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
CHEBI:50378
C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]

Specifications

104206-65-7
MFCD01752192
nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]
329.231
CHEBI:50378
95%
C14H10F3NO5
1g
OUBCNLGXQFSTLU-UHFFFAOYSA-N
2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
115355
329.23
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.