missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Morpholinoethoxy)phenyl]methylamine, 97%, Thermo Scientific™

Product Code. 10372245 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10372245 5 g 5g
10547493 250 mg 250mg
10442304 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10372245 Supplier Thermo Scientific Maybridge Supplier No. CC41913DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 540753-13-7
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD08060517
InChI Key QHAMTJRGPZWTSO-UHFFFAOYSA-N
Synonym 2-2-morpholinoethoxy phenyl methylamine, 2-2-morpholin-4-yl ethoxy phenyl methanamine, 2-2-morpholin-4-yl-ethoxy-benzylamine, 1-2-2-morpholin-4-yl ethoxy phenyl methanamine, 2-2-morpholin-4-ylethoxy phenyl methanamine, 2-2-morpholinoethoxy phenyl methanamine, 2-2-morpholin-4-ylethoxy phenyl methylamine
PubChem CID 7537544
IUPAC Name [2-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES C1COCCN1CCOC2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 236.31
Percent Purity 97%
Chemical Name or Material [2-(2-Morpholinoethoxy)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.