missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(4-chlorophenol), 95%

Product Code. 11460483
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460483 25 g 25g
11470483 100 g 100g
11480483 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460483 Supplier Thermo Scientific Alfa Aesar Supplier No. A12640.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-23-4 | C13H10Cl2O2 | 269.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-23-4
Molecular Formula C13H10Cl2O2
Molecular Weight (g/mol) 269.12
MDL Number MFCD00002322
InChI Key MDNWOSOZYLHTCG-UHFFFAOYSA-N
Synonym dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen
PubChem CID 3037
ChEBI CHEBI:34689
IUPAC Name 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O

Specifications

Melting Point 172°C to 176°C
Quantity 25 g
UN Number UN3077
Beilstein 1884514
Merck Index 14,3071
Formula Weight 269.13
Percent Purity 95%
Chemical Name or Material 2,2'-Methylenebis(4-chlorophenol)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.