missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethan-1-ol, 97%, Maybridge

Product Code. 10795993
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10795993 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795993 Supplier Thermo Scientific Maybridge Supplier No. BTB09707EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 443-48-1
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C6H9N3O3
Quantity 10 g
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
Molecular Weight (g/mol) 171.156
PubChem CID 4173
ChEBI CHEBI:6909
Formula Weight 171.16
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethan-1-ol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.