missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-4-thiazolyl)phenol, 97%, Thermo Scientific™

Product Code. 11365459
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365459 1 g 1g
11355459 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365459 Supplier Thermo Scientific Alfa Aesar Supplier No. H50431.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160241-65-6
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD00665949
InChI Key LYLPLPWQCLBGHW-UHFFFAOYSA-N
Synonym 2-2-methylthiazol-4-yl phenol, 2-2-methyl-1,3-thiazol-4-yl phenol, 2-2-methyl-4-thiazolyl phenol, acmc-1c1by, phenol,2-2-methyl-4-thiazolyl, 2-2-methyl-thiazol-4-yl-phenol
PubChem CID 18801453
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)phenol
SMILES CC1=NC(=CS1)C2=CC=CC=C2O

Specifications

Melting Point 61°C to 64°C
Quantity 1 g
Formula Weight 191.25
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-4-thiazolyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.