missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1H-imidazol-1-yl)benzoic acid, Tech., Thermo Scientific™

Product Code. 10255884
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255884 1 g Amber Glass Bottle 1g
10738055 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255884 Supplier Thermo Scientific Maybridge Supplier No. CC61201DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159589-71-6
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.21
MDL Number MFCD09065012
InChI Key PDOVVVXGESUKQF-UHFFFAOYSA-N
PubChem CID 24229636
IUPAC Name 2-(2-methylimidazol-1-yl)benzoic acid
SMILES CC1=NC=CN1C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-2-methyl-1h-imidazol-1-yl benzoic acid, 2-2-methylimidazol-1-yl benzoic acid, 2-2-methylimidazolyl benzoic acid, 2-2-methyl-1h-imidazole-1-yl benzoic acid, benzoic acid,2-2-methyl-1h-imidazol-1-yl, benzoic acid, 2-2-methyl-1h-imidazol-1-yl
Formula Weight 202.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.