missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 142.23 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction