missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Thermo Scientific™
£31.15 - £85.15
Chemical Identifiers
CAS | 121357-04-8 |
---|---|
Molecular Formula | C6H9NOS |
Molecular Weight (g/mol) | 143.204 |
InChI Key | REKVZAFNJAMAQL-UHFFFAOYSA-N |
Synonym | 2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole |
PubChem CID | 19876739 |
IUPAC Name | 2-(2-methyl-1,3-thiazol-4-yl)ethanol |
SMILES | CC1=NC(=CS1)CCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10225414
|
Thermo Scientific Maybridge
CC54609CB |
250 mg |
£31.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10001534
|
Thermo Scientific Maybridge
CC54609DA |
1 g |
£85.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
121357-04-8 | |
143.204 | |
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole | |
2-(2-methyl-1,3-thiazol-4-yl)ethanol |
C6H9NOS | |
REKVZAFNJAMAQL-UHFFFAOYSA-N | |
19876739 | |
CC1=NC(=CS1)CCO |
Specifications
121357-04-8 | |
C6H9NOS | |
REKVZAFNJAMAQL-UHFFFAOYSA-N | |
2-(2-methyl-1,3-thiazol-4-yl)ethanol | |
19876739 | |
2-(2-Methyl-1, 3-thiazol-4-yl)ethanol |
97% | |
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole | |
CC1=NC(=CS1)CCO | |
143.204 | |
143.21 |