Learn More
2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Maybridge
Brand: Maybridge CC54609DA
Code : 31
Additional Details : CAS Number : 121357-04-8 Weight : 0.05100kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
121357-04-8 | |
143.204 | |
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole | |
2-(2-methyl-1,3-thiazol-4-yl)ethanol |
C6H9NOS | |
REKVZAFNJAMAQL-UHFFFAOYSA-N | |
19876739 | |
CC1=NC(=CS1)CCO |
Specifications
2-(2-Methyl-1, 3-thiazol-4-yl)ethanol | |
121357-04-8 | |
C6H9NOS | |
REKVZAFNJAMAQL-UHFFFAOYSA-N | |
2-(2-methyl-1,3-thiazol-4-yl)ethanol | |
19876739 |
1g | |
97% | |
2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole | |
CC1=NC(=CS1)CCO | |
143.204 | |
143.21 |