missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10411884
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411884 250 mg Amber Glass Bottle 250mg
10182934 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411884 Supplier Thermo Scientific Maybridge Supplier No. CC68401CB
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 65032-66-8
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD09879980
InChI Key BNRSCIXYHUTATP-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl benzoic acid, benzoic acid,2-2-methyl-4-thiazolyl, 2-2-methylthiazol-4-yl benzoic acid, 2-2-methyl-thiazol-4-yl-benzoic acid, 4-2-carboxyphenyl-2-methyl-1,3-thiazole
PubChem CID 12384552
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)benzoic acid
SMILES CC1=NC(=CS1)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.