missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenoxy)ethylamine, 97%, Thermo Scientific™

Product Code. 11449380
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449380 1 g 1g
11439380 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449380 Supplier Thermo Scientific Alfa Aesar Supplier No. H50251.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(2-Methoxyphenoxy)ethylamine is used for its reaction with 4-(2,3-epoxypropoxy) carbazole for preparation of Carvedilol (I).

Solubility
Difficult to mix.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1836-62-0
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00235185
InChI Key CKJRKLKVCHMWLV-UHFFFAOYSA-N
Synonym 2-2-methoxyphenoxy ethylamine, 2-2-methoxyphenoxy ethanamine, 1-2-aminoethoxy-2-methoxybenzene, 2-2-methoxyphenoxy ethyl amine, 2-2-methoxy-phenoxy-ethylamine, ethanamine, 2-2-methoxyphenoxy, unii-20s0vx4qt9, 2-2'-methoxy phenoxyethylamine, 2-2-methoxyphenoxy ethan-1-amine
PubChem CID 1713005
IUPAC Name 2-(2-methoxyphenoxy)ethanamine
SMILES COC1=CC=CC=C1OCCN

Specifications

Density 1.11
Boiling Point 98°C (0.4 mmHg)
Odor Amine-like
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Difficult to mix.
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.