missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Iodophenyl-d4)-N,N-dimethylacetamide, TRC

Product Code. 30394998
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394998 50 mg -
30408040 100 mg 100mg
30400647 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394998 Supplier Toronto Research Chemicals Supplier No. TRCI71753750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217360-63-8
Molecular Formula C10H8D4INO
Molecular Weight (g/mol) 293.14
Synonym 6-Iodo-N,N-dimethyl-benzene-2,3,4,5-d4-acetamide
IUPAC Name N,N-dimethyl-2-(2,3,4,5-tetradeuterio-6-iodophenyl)acetamide
SMILES [2H]c1c([2H])c([2H])c(CC(=O)N(C)C)c(I)c1[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12INO/c1-12(2)10(13)7-8-5-3-4-6-9(8)11/h3-6H,7H2,1-2H3/i3D,4D,5D,6D
Formula Weight 293.0215
Chemical Name or Material 2-(2-Iodophenyl-d4)-N,N-dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.