missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Iminobis[N-[2-(2-chlorobenzoyl)-4-nitrophenyl]acetamide], TRC

Product Code. 30392176
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392176 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392176 Supplier Toronto Research Chemicals Supplier No. TRCI5868201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-Iminobis[N-[2-(2-chlorobenzoyl)-4-nitrophenyl]acetamide]
CAS 191085-33-3
Molecular Formula C30 H21 Cl2 N5 O8
Quantity 1 mg
Synonym N-[2-(2-Chlorobenzoyl)-4-nitrophenyl]-2-[[[[2-(2-chlorobenzoyl)-4-nitrophenyl]carbamoyl]methyl]amino]acetamide
InChI Formula InChI=1S/C30H21Cl2N5O8/c31-23-7-3-1-5-19(23)29(40)21-13-17(36(42)43)9-11-25(21)34-27(38)15-33-16-28(39)35-26-12-10-18(37(44)45)14-22(26)30(41)20-6-2-4-8-24(20)32/h1-14,33H,15-16H2,(H,34,38)(H,35,39)
SMILES [O-][N+](=O)c1ccc(NC(=O)CNCC(=O)Nc2ccc(cc2C(=O)c3ccccc3Cl)[N+](=O)[O-])c(c1)C(=O)c4ccccc4Cl
IUPAC Name 2-[[2-[2-(2-chlorobenzoyl)-4-nitroanilino]-2-oxoethyl]amino]-N-[2-(2-chlorobenzoyl)-4-nitrophenyl]acetamide
Molecular Weight (g/mol) 650.422
Formula Weight 649.077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.