missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Hydroxyethyl)octadecylamino]ethyl ester octadecanoic acid, TRC

Product Code. 30391642
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391642 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391642 Supplier Toronto Research Chemicals Supplier No. TRCH998250250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Hydroxyethyl)octadecylamino]ethyl ester octadecanoic acid
CAS 52497-24-2
Molecular Formula C40 H81 N O3
Quantity 250 mg
Synonym Octadecyl(2-hydroxyethyl)(2-stearoyloxyethyl)amine, Octadecyldiethanolamine monostearate, Stearyldiethanolamine monostearate, Stearyldiethanolamine stearate
InChI Formula InChI=1S/C40H81NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41(36-38-42)37-39-44-40(43)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h42H,3-39H2,1-2H3
SMILES O=C(OCCN(CCO)CCCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCC
IUPAC Name 2-[2-hydroxyethyl(octadecyl)amino]ethyl octadecanoate
Molecular Weight (g/mol) 624.076
Formula Weight 623.622
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.