missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Hydroxy-4,5-dimethoxybenzoyl)amino]-4-thiazolecarboxylic Acid (>90%), TRC

Product Code. 30397352
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397352 50 mg 50mg
30394991 25 mg -
30393106 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397352 Supplier Toronto Research Chemicals Supplier No. TRCH97278850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809272-85-2
Molecular Formula C13 H12 N2 O6 S
Molecular Weight (g/mol) 324.31
Synonym 4-Thiazolecarboxylic acid, 2-[(2-hydroxy-4,5-dimethoxybenzoyl)amino]- (ACI), 2-[(2-Hydroxy-4,5-dimethoxybenzoyl)amino]-4-thiazolecarboxylic acid (ACI), 2-(2-Hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylic Acid, 2-(2-Hydroxy-4,5-dimethoxybenzamido)-1,3-thiazole-4carboxylic Acid
IUPAC Name 2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxylic acid
SMILES O=C(C1=CSC(NC(C2=CC(OC)=C(OC)C=C2O)=O)=N1)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H12N2O6S/c1-20-9-3-6(8(16)4-10(9)21-2)11(17)15-13-14-7(5-22-13)12(18)19/h3-5,16H,1-2H3,(H,18,19)(H,14,15,17)
Formula Weight 324.04
Chemical Name or Material 2-[(2-Hydroxy-4,5-dimethoxybenzoyl)amino]-4-thiazolecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.