missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methylamine, ≥97%, Thermo Scientific™

Product Code. 10082414
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082414 250 mg Amber Glass Bottle 250mg
10728275 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082414 Supplier Thermo Scientific Maybridge Supplier No. CC46213CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 771573-25-2
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD06213664
InChI Key KUTREPQQCILHIM-UHFFFAOYSA-N
Synonym 2-2-furyl phenyl methylamine, 2-furan-2-yl phenyl methanamine, 1-2-furan-2-yl phenyl methanamine, 2-furan-2-yl-benzylamine
PubChem CID 18525791
IUPAC Name [2-(furan-2-yl)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [2-(2-Furyl)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.