missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)-1,3-diphenylimidazolidine, 96%, Thermo Scientific™

Product Code. 15494377
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494377 250 mg 250mg
15484377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494377 Supplier Thermo Scientific Alfa Aesar Supplier No. H31757.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94378-00-4
Molecular Formula C19H18N2O
Molecular Weight (g/mol) 290.366
MDL Number MFCD00225139
InChI Key GGKPIWBVFHMQMP-UHFFFAOYSA-N
Synonym 2-furan-2-yl-1,3-diphenylimidazolidine, 2-2-furyl-1,3-diphenylimidazolidine, imidazolidine, 2-2-furanyl-1,3-diphenyl, acmc-20lynj, 2-1,3-diphenylimidazolidin-2-yl furan, 2-furan-2-yl-1,3-diphenyl-imidazolidine
PubChem CID 680997
IUPAC Name 2-(furan-2-yl)-1,3-diphenylimidazolidine
SMILES C1CN(C(N1C2=CC=CC=C2)C3=CC=CO3)C4=CC=CC=C4

Specifications

Melting Point 129°C to 131°C
Quantity 250 mg
Formula Weight 290.37
Percent Purity 96%
Chemical Name or Material 2-(2-Furyl)-1,3-diphenylimidazolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.