missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furil dioxime, mixture of isomers, 97%

Product Code. 11416856
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416856 1 g 1g
11426856 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416856 Supplier Thermo Scientific Alfa Aesar Supplier No. B23972.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 522-27-0 | C10H8N2O4 | 220.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 522-27-0
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
MDL Number MFCD00075315
InChI Key RBOZTFPIXJBLPK-DSOJMZEYSA-N
Synonym alpha-furil dioxime, furildioxime, alpha-furyldioxime, di-2-furylglyoxime, bis 2-furyl glyoxime, di-2-furylethanedione dioxime, ethanedione, di-2-furanyl-, dioxime, glyoxime, di-2-furyl, furil, dioxime, 1z,2z-1,2-di furan-2-yl ethane-1,2-dione dioxime
PubChem CID 5359431
SMILES O\N=C(\C(=N/O)\C1=CC=CO1)/C1=CC=CO1

Specifications

Melting Point ∼165°C
Quantity 1 g
Beilstein 85894
Merck Index 14,4306
IUPAC Name N-[(1E)-1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Formula Weight 220.19
Percent Purity 97%
Chemical Name or Material 2,2'-Furil dioxime, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.