missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)pyrrolidine, 95%, Thermo Scientific Chemicals

Product Code. 15428268
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72216-04-7
Molecular Formula C10H12FN
Molecular Weight (g/mol) 165.211
MDL Number MFCD02663474
InChI Key NOJCVVKIBLHAGW-UHFFFAOYSA-N
Synonym 2-2-fluorophenyl pyrrolidine, 2-2-fluorophenyl-pyrrolidine, pyrrolidine,2-2-fluorophenyl, rs-2-2-fluoro-phenyl-pyrrolidine
PubChem CID 3613306
IUPAC Name 2-(2-fluorophenyl)pyrrolidine
SMILES C1CC(NC1)C2=CC=CC=C2F

Specifications

Color Colorless
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 165.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(2-Fluorophenyl)pyrrolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'