missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)ethanol, 99%

Product Code. 11325649
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325649 25 g 25g
11315649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325649 Supplier Thermo Scientific Alfa Aesar Supplier No. L19998.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 50919-06-7 | C8H9FO | 140.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50919-06-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002887
InChI Key HNIGZVZDWCTFPR-UHFFFAOYSA-N
Synonym 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol
PubChem CID 2733296
IUPAC Name 2-(2-fluorophenyl)ethanol
SMILES OCCC1=CC=CC=C1F

Specifications

Density 1.045
Boiling Point 72°C (1 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.51
Quantity 25 g
Formula Weight 140.16
Percent Purity 99%
Chemical Name or Material 2-(2-Fluorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.