missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, TRC

Product Code. 30402696
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402696 250 mg
30403339 500 mg
30398693 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402696 Supplier Toronto Research Chemicals Supplier No. TRCF595688250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1165936-00-4
Molecular Formula C13H18BFO3
Molecular Weight (g/mol) 252.09
Synonym 2-Fluoro-3-methoxyphenylboronic Acid Pinacol Ester
IUPAC Name 2-(2-fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES FC=1C(OC)=CC=CC1B2OC(C)(C)C(O2)(C)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H18BFO3/c1-12(2)13(3,4)18-14(17-12)9-7-6-8-10(16-5)11(9)15/h6-8H,1-5H3
Formula Weight 252.13
Chemical Name or Material 2-(2-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.