missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluoro[1,1'-biphenyl]-4-yl)-2,3-dimethylbutanedioic Acid Dimethyl Ester, TRC

Product Code. 30402750
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402750 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402750 Supplier Toronto Research Chemicals Supplier No. TRCF588768100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2-Fluoro[1,1'-biphenyl]-4-yl)-2,3-dimethylbutanedioic Acid Dimethyl Ester
CAS 42771-81-3
Molecular Formula C15 H16 Cl N3 O4 . C H4 O3 S
Quantity 100 mg
Synonym Flurbiprofen Impurity 1
InChI Formula InChI=1S/C15H16ClN3O4.CH4O3S/c1-21-8-3-4-10-11(5-8)19-14(13(16)18-10)23-9-6-12(17-7-9)15(20)22-2;1-5(2,3)4/h3-5,9,12,17H,6-7H2,1-2H3;1H3,(H,2,3,4)/t9-,12+;/m1./s1
SMILES COC(=O)[C@@H]1C[C@H](CN1)Oc2nc3cc(OC)ccc3nc2Cl.CS(=O)(=O)O
IUPAC Name methanesulfonic acid;methyl (2S,4R)-4-(3-chloro-7-methoxyquinoxalin-2-yl)oxypyrrolidine-2-carboxylate
Molecular Weight (g/mol) 433.864
Formula Weight 433.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.