missing translation for 'onlineSavingsMsg'
Learn More

2,2'-(Ethylenedioxy)bis(ethylamine), 97+%

Product Code. 11400011
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11400011

Brand: Thermo Scientific Alfa Aesar 044759.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 929-59-9 | C6H16N2O2 | 148.206 g/mol

2,2'-(Ethylenedioxy)bis(ethylamine) is used to linker to form conjugates of manganese-doped zinc sulfide nanoparticles with folic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-59-9
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.206
MDL Number MFCD00040474
InChI Key IWBOPFCKHIJFMS-UHFFFAOYSA-N
Synonym 1,8-diamino-3,6-dioxaoctane, 1,2-bis 2-aminoethoxy ethane, 2,2'-ethylenedioxy bis ethylamine, unii-r41zt4uf34, 2,2'-ethane-1,2-diylbis oxy diethanamine, 3,6-dioxaoctamethylenediamine, 2-2-2-aminoethoxy ethoxy ethanamine, 3,6-dioxaoctane-1,8-diamine, ethylene glycol bis 2-aminoethyl ether
PubChem CID 70248
IUPAC Name 2-[2-(2-aminoethoxy)ethoxy]ethanamine
SMILES C(COCCOCCN)N

Specifications

Density 1.01 g/mL
Boiling Point 105°C to 109°C (6 mmHg)
Flash Point 121°C (250°F)
Linear Formula C6H16N2O2
Refractive Index 1.461
Quantity 50 g
UN Number UN2735
Beilstein 1739187
Sensitivity Hygroscopic
Solubility Information Soluble in 0.4 parts water.
Formula Weight 148.2
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2,2'- (Ethylenedioxy)bis (ethylamine)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.