missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyphenoxy)ethanamine Hydrochloride, TRC

Product Code. 30369694
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369694 500 mg -
30381296 100 mg -
30384183 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369694 Supplier Toronto Research Chemicals Supplier No. TRCE892885500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1051368-80-9
Molecular Formula C10 H15 N O2 . Cl H
Molecular Weight (g/mol) 217.69
Synonym Ethanamine, 2-(2-ethoxyphenoxy)-, hydrochloride (1:1), 2-(2-Ethoxyphenoxy)ethylamine hydrochloride, 2-(o-Ethoxyphenoxy)ethylamine hydrochloride, β-(o-Ethoxyphenoxy)ethylamine hydrochloride
IUPAC Name 2-(2-ethoxyphenoxy)ethanamine;hydrochloride
SMILES Cl.CCOc1ccccc1OCCN

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H15NO2.ClH/c1-2-12-9-5-3-4-6-10(9)13-8-7-11;/h3-6H,2,7-8,11H2,1H3;1H
Formula Weight 217.087
Chemical Name or Material 2-(2-Ethoxyphenoxy)ethanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.