missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate

Product Code. 11318215
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318215 50 g 50g
11308215 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318215 Supplier Thermo Scientific Alfa Aesar Supplier No. L08522.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54550-36-6 | C6H13BrO2 | 197.072 g/mol

2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

Solubility
Not miscible with water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54550-36-6
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.072
MDL Number MFCD00051920
InChI Key UEDIWIFQWRXXJG-UHFFFAOYSA-N
Synonym 1-bromo-2-2-ethoxyethoxy ethane, 2-2-ethoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxyethane, 1-bromo-2-2-ethoxy-ethoxy-ethane, acmc-1axh5, 1-bromo-3,6-dioxaoctane, 1-2-ethoxyethoxy-2-bromoethane, 1-ethoxy-2-2-bromoethoxy ethane, 1-2-bromoethoxy-2-ethoxy-ethane, 1-bromo 2-2-ethoxyethoxy-ethane
PubChem CID 7015324
IUPAC Name 1-(2-bromoethoxy)-2-ethoxyethane
SMILES CCOCCOCCBr

Specifications

Density 1.299
Boiling Point 86°C to 88°C (12 mmHg)
Refractive Index 1.453
Quantity 50 g
Beilstein 1736603
Solubility Information Not miscible with water.
Formula Weight 197.08
Percent Purity 98%
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl bromide, stabilized with potassium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.