missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethyl acetate, 99%

Product Code. 10753894 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10753894 2.5 L Glass bottle 2.5 L
10277730 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753894 Supplier Thermo Scientific Supplier No. 257420025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-15-2 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Acidity 0.2 mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Color Beige to White
Density 1.0100g/mL
Boiling Point 210.0°C to 220.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)OCH2CH2OCH2CH2OC2H5
Refractive Index 1.4206 to 1.4226
Quantity 2.5 L
Residue after Evaporation 0.2% max.
Merck Index 15, 3134
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, most oils
Viscosity 2.8 mPa.s (20°C)
Formula Weight 176.2
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl acetate
Show More Show Less
compliance-icons
Produktidentifikator
  • 2-(2-Ethoxyethoxy)ethyl acetate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
Gefahrenhinweis
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.