missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiobisbenzoic Acid, TRC

Product Code. 30388028
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388028 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388028 Supplier Toronto Research Chemicals Supplier No. TRCD49399010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-Disulfanediyldibenzoic acid
CAS 119-80-2
Molecular Formula C14H10O4S2
Quantity 10 g
Synonym 2,2'-Dithiodibenzoic Acid, 2,2'-Dithiobis[benzoic acid], o,o’-Dithiodibenzoic acid, 2-[(2-Carboxyphenyl)disulfanyl]benzoic Acid, Bis(2-carboxyphenyl) Disulfide, Bis(o-carboxylphenyl) Disulfide, Diphenyl Disulfide-2,2'-dicarboxylic acid, Dithiosalicylic acid, NSC 213, NSC 405662, NSC 5346, NSC 633230
InChI Formula InChI=1S/C14H10O4S2/c15-13(16)9-5-1-3-7-11(9)19-20-12-8-4-2-6-10(12)14(17)18/h1-8H,(H,15,16)(H,17,18)
SMILES OC(=O)c1ccccc1SSc2ccccc2C(=O)O
IUPAC Name 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
Molecular Weight (g/mol) 306.4
Formula Weight 306.0021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.