missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl N,N'-dioxide, 98%

Product Code. 10760791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10760791 1 g Glass bottle 1g
10198133 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10760791 Supplier Thermo Scientific Acros Supplier No. 319610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7275-43-6 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7275-43-6
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00065169
InChI Key FERMVCULDZOVOJ-KTKRTIGZSA-N
Synonym 2,2'-bipyridine 1,1'-dioxide, 2,2'-dipyridyl n,n'-dioxide, 2,2'-bipyridine n,n'-dioxide, 2,2'-dipyridyl-n,n'-dioxide
PubChem CID 6005843
SMILES [O-]N1C=CC=C\C1=C1/C=CC=C[N+]1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 296°C to 298°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name (2E)-2-(1-oxidopyridin-2-ylidene)pyridin-1-ium 1-oxide
Formula Weight 188.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 2'-Dipyridyl N, N'-dioxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.