missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dipyridyl disulfide, 98%

Product Code. 11489003
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11489003 25 g 25g
11479003 5 g 5g
11499003 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11489003

Brand: Thermo Scientific Alfa Aesar A11118.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2127-03-9 | C10H8N2S2 | 220.31 g/mol

2,2'-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.

Notes
Incompatible with oxidizing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2127-03-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.31
MDL Number MFCD00006287
InChI Key HAXFWIACAGNFHA-UHFFFAOYSA-N
Synonym 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
PubChem CID 65093
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
SMILES C1=CC=NC(=C1)SSC2=CC=CC=N2

Specifications

Melting Point 56°C to 59°C
Flash Point 210°C (410°F)
Assay Percent Range 98%
Quantity 25 g
Beilstein 154629
Solubility Information Soluble in water,benzene,tetrahydrofuran,dichloromethane,acetonitrile and dimethyformamide.
Formula Weight 220.32
Percent Purity 98%
Chemical Name or Material 2,2'-Dipyridyl disulfide
compliance-icons
Product Identifier
  • 2,2'-Dipyridyl disulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,2-Dithiodipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.