missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(Dipropylamino)ethyl]-6-nitrophenyl Acetic Acid Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[2-(Dipropylamino)ethyl]-6-nitrophenyl acetic acid hydrochloride |
| CAS | 91374-25-3 |
| Molecular Formula | C16 H24 N2 O4 . Cl H |
| Quantity | 250 mg |
| Synonym | Benzeneacetic acid, 2-[2-(dipropylamino)ethyl]-6-nitro-, hydrochloride (1:1), Benzeneacetic acid, 2-[2-(dipropylamino)ethyl]-6-nitro-, monohydrochloride (9CI), 6-[2-(Dipropylamino)ethyl]-2-nitrophenylacetic acid hydrochloride, [2-Nitro-6-(2-dipropylaminoethyl)phenyl]acetic acid hydrochloride |
| InChI Formula | InChI=1S/C16H24N2O4.ClH/c1-3-9-17(10-4-2)11-8-13-6-5-7-15(18(21)22)14(13)12-16(19)20;/h5-7H,3-4,8-12H2,1-2H3,(H,19,20);1H |
| SMILES | Cl.CCCN(CCC)CCc1cccc(c1CC(=O)O)[N+](=O)[O-] |
| IUPAC Name | 2-[2-[2-(dipropylamino)ethyl]-6-nitrophenyl]acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 344.83 |
| Formula Weight | 344.1503 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?